Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Mercaptoethylamine-HCl

Catalog No. PI20408 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 x 6 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20408 6 x 6 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20408 Supplier Thermo Scientific™ Supplier No. 20408
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pure, crystalline 2-aminoethanethiol (2-MEA-HCl, also called cysteamine-HCl), CAS 156-57-0; selectively reduces antibody hinge-region disulfide bonds.

Thermo Scientific™ Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.

M.W. 113.61
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store desiccated at 4°C. Product is shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 6 x 6 mg
Product Type Mercaptoethylamine Hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.