Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ 96-Well Polystyrene Plates, White Opaque

Catalog No. PI15042 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15042 25 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15042 Supplier Thermo Scientific™ Supplier No. 15042
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.

Thermo Scientific™ Pierce 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.

Thermo Scientific Pierce White Opaque 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics. These microplates have a corner notch for use in standard plate readers.

Features of White Opaque 96-Well Microplates:

  • White opaque plates for fluorescent or chemiluminescent assays
  • Treated to provide protein coating capacity
  • High antibody-binding surface improves the overall performance in immunoassay applications
  • Plates are certified for performance, having consistently low coefficients of variation among wells

Plate Properties:

  • Antibody binding capacity: High, 400ng IgG/cm2 (1 cm2 = 100 μL volume)
  • Binding interaction: hydrophobic-ionic
  • Well-to-well coefficient of variation: CV <10%
  • Well shape (bottom): Flat
  • Maximum well volume: 360 μL

Related Products

  • Pierce™ 96-Well Polystyrene Plates, Corner Notch (Cat. No. 15041)
TRUSTED_SUSTAINABILITY

Specifications

Binding Property Adsorption, Hydrophilic
Description Pierce 96-Well Polystyrene Plates, White Opaque
Detection Method Colorimetric
Packaging 25 Plate
Product Line Pierce
Color White
Product Type Well Plate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Polystyrene
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 25 plates
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.