Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Chromatin Prep Module

Catalog No. PI26158 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26158 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26158 Supplier Thermo Scientific™ Supplier No. 26158
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chromatin immunoprecipitation (chromatin IP, ChIP) assay kit containing all reagents, agarose affinity beads, and a simple, fast, reliable protocol.

Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.

This chromatin IP kit contains sufficient reagents to perform 30 ChIP assays with appropriate controls using an optimized protocol and a convenient spin-column format. The kit includes a complete Chromatin Prep Module (also available separately) to efficiently extract and solubilize crosslinked DNA-protein interaction complexes from cells, plus an immunoprecipitation module that contains high-capacity, ChIP-grade Protein A/G agarose.

Highlights:

  • Simple and streamlined ChIP assay protocol (< 8 hours)
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-capacity Protein A/G agarose resin
  • Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
  • Fast and reproducible spin-column format; columns and collection tubes included
  • High-recovery DNA purification
  • Choose a complete ChIP assay kit or the separate chromatin preparation module

Includes:

Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Chromatin Prep Module
Content And Storage Sufficient For: 30 chromatin preparations for ChIP assays
• Membrane Extraction Buffer, 10 mL
• Nuclear Extraction Buffer, 10 mL
• MNase Digestion Buffer, 5 mL
• MNase Stop Solution, 0.5 mL
• Halt Protease and Phosphatase Inhibitor Cocktail EDTA-free (100X), 4 x 90 μL
• Sodium chloride (5M), 3 mL
• Glycine solution (10X), 15 mL
• PBS (20X), 15 mL
• Micrococcal Nuclease (ChIP Grade) (10 U/μL) (store at -20°C)
• Proteinase K (20 mg/mL), 0.25 mL (store at -20°C)
• DTT, Lyophilized, 1 vial

Unless otherwise indicated, store all components at 4°C.
Product Line Pierce
Quantity 30 reactions
Type Chromatin Isolation Kit
Sufficient For 30 Reactions
Format Kit
Binding Capacity 2 x 106 Cells/Reaction
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.