Learn More
Description
Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
Highlights:
- Simple and streamlined ChIP assay protocol (< 8 hours)
- Highly efficient isolation and lysis of nuclei
- Easy and reproducible enzymatic digestion
- Low-background and high-capacity Protein A/G agarose resin
- Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
- Fast and reproducible spin-column format; columns and collection tubes included
- High-recovery DNA purification
- Choose a complete ChIP assay kit or the separate chromatin preparation module
Includes:
Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C |
| Form | Liquid Suspension |
| Product Line | Pierce |
| Quantity | 10 mL |
| Target | IgG Antibody |
| Type | Agarose |
| Stationary Phase | Protein A/G |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.