Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ammonium Persulfate

Catalog No. PI17874 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17874 4 x 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17874 Supplier Thermo Scientific™ Supplier No. 17874
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Catalyze acrylamide and bisacrylamide polymerization when preparing polyacrylamide gels for electrophoresis with APS oxidizing agent.

Thermo Scientific™ Pierce Ammonium Persulfate (APS) is an oxidizing agent that is used with TEMED to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.

Ammonium persulfate (APS) is an oxidizing agent that is often used with tetramethylethylenediamine (TEMED, Part No. 17919) to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.

Highlights:

  • Name: Ammonium persulfate (APS)
  • Formula: (NH4)2S2O8
  • CAS number: 7727-54-0
  • Molecular weight: 228.2
  • Purity: ≥98.0%
  • Insolubles: ≤0.005%

Compatible with:

TEMED, Part No. 17919

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store below 23°C.
Form Powder
Quantity 4 x 25 g
Product Type Ammonium Persulfate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.