Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Anti-DYKDDDDK Affinity Resin

Catalog No. A40004562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 mL
5 Columns
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40004562 5 Columns
PIA36803 10 mL
PIA36801 2 mL
PIA36804 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A40004562 Supplier Thermo Scientific™ Supplier No. A40004562
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells.

Thermo Scientific Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to UltraLink Biosupport, which is a rigid, hydrophilic, highly crosslinked, copolymeric, and porous resin with high coupling capacity.

For protein purification, resin is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or C-terminus. Following capture of DYKDDDDK-tagged proteins, non-specifically bound proteins can be washed away before dissociating bound target proteins with elution buffer.

Features of Pierce Anti-DYKDDDDK Affinity Resin

  • Specific—unique base beads and highly specific antibody minimize off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Flexible—available in multiple formats including bulk loose resin and spin columns

Characteristics of Pierce Anti-DYKDDDDK Affinity Resin

  • Composition: anti-DYKDDDDK antibody covalently attached to UltraLink Biosupport, a highly crosslinked rigid support
  • Particle size: 50–80 μm
  • pH stability: 1–13
  • Operating temperature: 2–30°C; do not freeze
  • Particle concentration: 50% slurry in phosphate buffered saline, 0.02% sodium azide, pH 7.5
  • Binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)

Available formats

  • Resin slurries—2 mL, 10 mL, and 100 mL of 50% slurry in PBS with 0.02% sodium azide, pH 7.5
  • Spin columns—10 mL spin columns containing 3 mL of settled resin, supplied at 6 mL 50% slurry
TRUSTED_SUSTAINABILITY

Specifications

Product Type Affinity Resin
Quantity 5 Columns
Particle Size 50 to 80 μm
Sufficient For ≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)
Capacity 10 mL
Stationary Phase Anti-DYKDDDDK monoclonal antibodies
Shipping Condition Wet Ice
Form Liquid Suspension

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.