Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Anti-GST Coated Plates, Clear, 8-Well Strip

Catalog No. PI15144 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15144 5 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15144 Supplier Thermo Scientific™ Supplier No. 15144
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect glutathione S-transferase (GST)-tagged proteins with these ready-to-use pre-blocked, mouse monoclonal anti-GST antibody coated polystyrene microplates.

Thermo Scientific™ Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.

Additional Information and Online Ordering

Unlike glutathione coated plates, these anti-GST plates effectively bind both native and denatured forms of GST. In most cases, pre-purification of cell lysates is not necessary before screening and analysis of recombinant GST-tagged protein expression by ELISA using the plates.

Highlights:

  • Antibody-based – plates are coated with mouse monoclonal anti-GST antibody

  • Sensitive – detects GST fusion proteins down to 7.5ng/well

  • Flexibility – binds native or denatured forms of GST; an alternative to glutathione-coated plates

  • Ready to use – pre-blocked with SuperBlock Blocking Buffer

  • Compatibility – Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding

  • Superior performance – improved signal-to-noise ratio relative to competitor plates

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Affinity
Content And Storage Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature.
Detection Method Colorimetric
Packaging 5 Plate
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment Antibody
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.