Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Biotin Coated Plates, Clear, 8-Well Strip

Catalog No. PI15151 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15151 5 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15151 Supplier Thermo Scientific™ Supplier No. 15151
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capture NeutrAvidin protein, streptavidin, avidin or any other biotin-binding protein with these biotin-coated plates.

Thermo Scientific™ Pierce Biotin Coated Plates can be used in any immunoassay with Thermo Scientific™ NeutrAvidin Biotin-Binding Protein, streptavidin, avidin or other biotin-binding proteins.

Highlights:

  • Biotin group accessible for binding avidin, streptavidin or NeutrAvidin™ Biotin-Binding Protein
  • Pre-blocked to reduce nonspecific binding
  • Strip well plate format for the ultimate in convenience
  • Coat volume: 200μL
  • Blocking volume: 300μL
TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Affinity
Content And Storage Store desiccated at 4°C.
Detection Method Colorimetric
Packaging 5 Plate
Product Line Pierce
Color Clear
Product Type Coated Strip Plate
Material Polystyrene
Volume (Metric) Well 200 μL
No. of Wells 96
Surface Treatment Biotin
Plate Blocking Proprietary
Quantity 5 plates
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.