Learn More
Description
Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.
Highlights:
Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining
Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)
Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods
Specifications
Specifications
| Conjugate | Biotin |
| Description | Pierce Biotin |
| Quantity | 1 g |
| Content And Storage | Store in a cool, dry area, protected from direct sunlight. |
| Reactive Moiety | Carboxylic Acid |
| Solubility | DMSO (Dimethylsulfoxide), Water, DMF (Dimethylformamide) |
| Form | Powder |
| Product Line | Pierce |
| Product Type | Biotin (Unconjugated) |
| Label or Dye | Biotin & Analog |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.