Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Bovine Serum Albumin, Biotinylated

Catalog No. PI29130 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI29130 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI29130 Supplier Thermo Scientific™ Supplier No. 29130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified biotin and biotinylated BSA, HRP, AP or FITC for use as controls or to amplify signal in IHC via the avidin-biotin complex (ABC).

Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.

Biotinylated HRP and biotinylated AP are most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method. Biotinylated Fluorescein (FITC) cannot be used to polymerize and amplify the signal to the same degree as biotinylated enzymes because each fluor molecule is tagged with only one biotin molecule. However, the fluorescent variant of traditional ABC staining is possible with this unique molecule.

Highlights:

  • Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining

  • Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)

  • Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Pierce Bovine Serum Albumin, Biotinylated
Conjugate Biotin
Content And Storage Store between 2°C and 8°C.
Label Type Biotin & Analogs
Form Solid
Product Line Pierce
Protein BSA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.