Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Biotinylated Protein Interaction Pull-Down Kit
GSA_VA

Catalog No. PI21115 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21115 25 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21115 Supplier Thermo Scientific™ Supplier No. 21115
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Easily capture and affinity-purify binding partners of any biotin-labeled protein or other ligand using these streptavidin agarose beads and complete reagent set.

Thermo Scientific™ Pierce Biotinylated Protein Interaction Pull-Down Kit contains the necessary components to capture and purify interactors of a biotin-labeled protein or ligand.

The kit makes it possible to perform an efficient pull-down assay with high protein recovery and reproducibility. This protocol uses immobilized streptavidin to capture a biotinylated protein and a spin column format that enables complete gel retention during each capture, wash and elution step. When the biotinylated bait protein is immobilized, it can withstand stringent washes and remains bound during prey protein elution, which results in an isolated prey protein without bait protein contamination. This technique is ideal when using native or recombinant proteins that are not expressed with a fusion tag. Additionally, prey protein is isolated from the immobilized bait protein using native binding conditions and nondenaturing elution conditions. The Biotinylated Protein Interaction Pull-Down Kit is designed to teach the method to first-time users and to increase ease-of-use and convenience for experienced researchers.

Highlights:

  • Biotin pull-down – purifies protein interactors of any biotinylated protein or biotin-tagged ligand

  • Complete kit – provides all components and detailed protocol for purifying protein:protein interactions

  • No special equipment needed – uses common laboratory equipment and reagents (e.g., microcentrifuge)

  • Spin column efficiency – microcentrifuge spin columns facilitate microcentrifuge spin columns facilitate simple and efficient manipulation of streptavidin agarose beads, including simple processing of multiple samples

Includes:

Streptavidin agarose resin, TBS pack, biotin blocking buffer, wash buffer, elution buffer, spin columns and collection tubes

Recommended for:

Discover a new protein interaction complex using a biotinylated protein as the bait to capture a putative binding partner from a cell lysate or other sample; Confirm expression of a known protein interaction target in a cell lysate; Determine the ability of two purified proteins to interact (bind) in various buffer conditions; Extract protein:protein interaction information from in vitro transcription/ translation lysates

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: At least 25 pull-down reactions, each using 25 μL of resin
• Streptavidin Agarose Resin, 1.5 mL
• TBS Pack (makes 500mL), 1 pack
• Biotin Blocking Solution, 15 mL
• Wash Buffer (pH 5.0), 100 mL
• Elution Buffer (pH 2.8), 50 mL
• Pierce Spin Columns and Accessories, 27 columns
• Microcentrifuge Collection Tubes, 200 tubes

Store at 4°C
Assay Affinity Purification
For Use With (Equipment) Microcentrifuge
Product Line Pierce
Quantity 25 reactions kit
For Use With (Application) Pull-Down (Biotin-Based)
Target Not Target-Specific
Type Pull-Down Kit
Sufficient For 25 Reactions
Format Kit
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.