Learn More
Thermo Scientific™ Pierce™ Bovine Gamma Globulin Standard Ampules, 2 mg/mL
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce BGG Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.
Highlights:
Antibody standard – best reference standard for immunoglobulin quantitation in colorimetric protein assays
Bradford standard – best general protein standard in coomassie-based (Bradford) protein assays
Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL, ensuring excellent lot-to-lot consistency
Convenient – choose a package of 1mL ampules or a complete set containing seven ready-to-use dilutions
Includes:
2mg/mL BGG solution (Part No. 23212) or 7-solution set of different concentrations (Part No. 23213)
Recommended for:
Protein assay immunoglobulin quantitation standard; Antibody recovery control for desalting and other column procedures; Loading control for SDS-PAGE; General calibration of spectrophotometer UV-lamp (A280)
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Description | Pierce Bovine Gamma Globulin Standard Ampules, 2mg/mL |
| Assay | Protein Standards |
| Product Line | Pierce |
| Product Type | Protein Quantitation Assay |
| Specificity | Not Target-Specific |
| For Use With (Application) | Internal Standard |
| Quantity | 10 x 1 mL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.