Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CHAPS Detergent (3-((3-cholamidopropyl) dimethylammonio)-1-propanesulfonate)
GSA_VA

Catalog No. p-4530894
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28299 100 g
PI28300 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI28299 Supplier Thermo Scientific™ Supplier No. 28299
Only null left
Add to Cart
Add to Cart

Non-denaturing Zwitterionic detergents that disrupt nonspecific protein interactions with less protein aggregation than nonionic detergents.

Thermo Scientific™ CHAPS is a zwitterionic detergent that is especially well suited for protecting the native state of proteins.

CHAPS is a sulfobetaine derivatives of cholic acid. This zwitterionic detergent is useful for membrane protein solubilization when it is important to maintain protein activity. CHAPS detergent is soluble over a wide range of pH (2 to 12) and are easily removed from solution by dialysis because of their high critical micelle concentration (CMC).

Highlights:

  • Versatile zwitterionic detergents
  • Provide mild but effective lysis of cultured mammalian cells
  • Nondenaturing and generally do not inactivate protein functions
  • Can be removed from solution by dialysis
  • Commonly used for isoelectric focusing (IEF) and 2D electrophoresis

Properties of CHAPS detergent:

  • Chemical Name: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
  • Molecular Weight: 614.88
  • Detergent Class: Zwitterionic
  • Aggregation Number: 10
  • Micelle Molecular Weight: 6149
  • Critical Micelle Concentration (CMC): 8 to 10mM (0.4920 to 0.6150%, w/v)
  • Cloud Point: ≥ 100°C
  • Dialyzable: Yes

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store at Room Temperature
Form Powder
Reagent Type Detergent (Pure)
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.