Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Compat-Able™ Bradford Plus Protein Assay Kit

Catalog No. PI23239 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23239 1.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23239 Supplier Thermo Scientific™ Supplier No. 23239
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.

Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.

The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.

Highlights:

  • Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer

  • Simple and fast – prepare samples in less than 10 minutes with four simple steps

  • Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results

  • Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs

  • Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures

Includes:

Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: Treating up to 500 protein assay samples and performing 630 tube assays or 3160 microplate assays
• Compat-Able Reagent 1, 250 mL
• Compat-Able Reagent 2, 250 mL
• Pierce Coomassie Plus Assay Reagent, 950 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL
Description Compat-Able Coomassie Plus Protein Assay Kit
Assay Bradford Assay
Detection Method Colorimetric
Product Line Compat-Able
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 630 Tube Assays or 3160 Microplate Assays
For Use With (Application) Absorbance, Solution-based Detection, Incompatibility Elimination
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 1.5 L
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.