Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Glycoprotein Isolation Kit, WGA

Catalog No. PI89805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89805 10 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89805 Supplier Thermo Scientific™ Supplier No. 89805
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Purify glycoproteins from serum and cell lysates using affinity resins or kits with concanavilin A (ConA) or wheat germ agglutinin (WGA) lectins.

Thermo Scientific™ Pierce Glycoprotein Isolation Kits use ConA and WGA lectin affinity resins and optimized buffers to efficiently isolate glycoproteins from serum, tissue and cultured cell lysates, thus enabling downstream analysis.

These two kits use lectins concanavilin A (ConA) and wheat germ agglutinin (WGA), respectively, as ligands to bind specific classes of glycoproteins from complex protein samples. ConA lectin recognizes alpha-linked mannose and terminal glucose residues, while WGA lectin selectively binds to N-Acetyl glucosamine (GlcNAc) groups and to sialic acid. The kits include ConA or WGA beaded agarose resin, optimized binding/wash and elution buffers, and easy-to-use microcentrifuge spin columns. The kits have been used to successfully isolate glycoproteins from serum as well as HeLa and CHO cell lysates in approximately 50 minutes.

Glycosylation is a post-translational modification that plays an important role in biological functions, including immune regulation, inflammation, cell-to-cell adhesion and cell signaling. Asparagine (N-linked) and serine/threonine residues (O-linked) are glycosylated during passage through the endoplasmic reticulum and golgi apparatus in eukaryotic and prokaryotic (i.e., Archaea and Eubacteria) systems. Glycoconjugates are important for immune regulation, inflammation, cell-to-cell adhesion and contact inhibition, cell signaling, protection against proteolytic degradation, and other biological processes.

Highlights:

  • High recovery – equivalent or greater glycoprotein recovery than the kits and lectin resins of other suppliers

  • Fast – glycoprotein purification in less than one hour

  • Versatile – isolate glycoproteins from various sample types; e.g., human serum and cell lysate

  • Robust – lectin does not leach from resin when processing sample

  • Convenient – complete kits include lectin resins and spin columns with all necessary reagents

  • Compatible with Bradford-based protein assays – dialysis or protein precipitation of recovered glycoproteins is not required prior to protein assay

Includes:

ConA or WGA beaded agarose resin (1.1mL), binding/wash buffer, elution buffer, and microcentrifuge spin columns and accessories.

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Glycoprotein Isolation Kit, WGA
Content And Storage Sufficient For: Isolating glycoproteins with strong affinity for WGA from 10 samples of up to 640 μL (1 to 1.5 mg total protein) each
• WGA Lectin Resin, 1.1 mL
• Binding/Wash Buffer (5X), 6.5 mL
• Elution Buffer, 5 mL
• Spin Columns, 10 columns
• Collection Tubes, 20 tubes

Store at 4°C.
Product Line Pierce
Quantity 10 reactions kit
Target Glycoprotein
Type Glycoprotein Isolation Kit
Stationary Phase Lectin
Sufficient For 10 Reactions
Format Kit, Microcentrifuge, Spin Column
Column Type Affinity, Agarose Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.