Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Control Agarose Resin

Catalog No. PI26150 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26150 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26150 Supplier Thermo Scientific™ Supplier No. 26150
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Unmodified beaded agarose resin for pre-clearing protein immunoprecipitation samples to reduce background signal and nonspecific binding in IP assays.

Thermo Scientific™ resin is effective for depleting proteins and other sample components that bind to agarose nonspecifically, which reduces background signal in IP reactions

When used as a negative control, these beads enable specific and nonspecific binding interactions to be distinguished.

Highlights:

  • Crosslinked 4% beaded agarose (CL4B) is the same base support used in various Thermo Scientific Pierce IP and co-IP kits, unmodified and non-activated
  • 10mL package size is more appropriate for experimental needs
  • Supplied as a 50% slurry in 20% ethanol (pipetting 1mL of slurry delivers 0.5mL of settled resin): wash in buffer and the beads are ready to use
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C.
Form Liquid Suspension
Product Line Pierce
Quantity 10 mL
Type Resin
Format Bottle
Column Type Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.