Learn More
Thermo Scientific™ Pierce™ Crossflow Diafiltration Reservoir, 500 mL
Catalog No. FERA57935 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
FERA57935 Each
1 options
Catalog No. FERA57935 Supplier Thermo Scientific™ Supplier No. A57935
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart
Pierce Crossflow Diafiltration Reservoir, for continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
The Pierce Crossflow Diafiltration Reservoir allows for continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
• For continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Plastic
Sterility Non-sterile
Type Self-standing Reagent Reservoir with Lid
Includes 3 ports
Quantity Each
Autoclavable Not Autoclavable
Disposable Reusable
Bottom Shape Flat