Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Sodium meta-Periodate

Catalog No. PI20504 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20504 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20504 Supplier Thermo Scientific™ Supplier No. 20504
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Gently oxidize glycoprotein carbohydrate sugars to create reactive aldehydes (carbonyls) for crosslinking and immobilization using NaIO4 (CAS 7790-28-5).

Thermo Scientific™ Pierce Sodium meta-Periodate is a gentle oxidizing agent that effectively cleaves cis-diols in carbohydrate sugars to create reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.

Highlights:

  • Capable of the oxidation of cis-diols to aldehydes
  • ACS reagent grade
  • White, crystalline, free-flowing
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at room temperature.
Label or Dye Aldehyde Group
Label Type Conjugating Groups
Product Line Pierce
Form Powder
Quantity 25 g
Product Type Sodium Meta-Periodate
What would be the best way to crosslink an antibody with horseradish peroxidase (‘HRP’ unmodified)?

A commonly used method for conjugating HRP to antibodies is by activation of the HRP using sodium meta-periodate (Cat no. 20504) to generate aldehydes. After the reaction, the HRP must be dialyzed to remove the meta-periodate; then the aldehydes on the HRP will react directly to the amine groups on the antibody. Alternatively, we offer activated HRP (Cat. No. 31488) as an alternative to avoid the periodate oxidation step to modify the HRP. 

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.