Learn More
Description
Assay regulatory element transcriptional activity in mammalian cell culture media and whole cell lysate with this highly sensitive flash-based luciferase kit.
The Thermo Scientific Pierce Cypridina Luciferase Flash Assay Kit provides researchers with a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.
Features of the Cypridina Luciferase Flash Assay Kit:
- Sensitive—high sensitivity allows utilization of smaller numbers of cells
- Cost effective—sensitive assays result in decreased reagent consumption
- Secreted luciferase—allows real-time assays and kinetic studies without destroying cells
- Ideal for co-transfection—uses a different substrate than Renilla, Gaussia, or firefly luciferases
- Compatible—assay reagents compatible with other Cypridina or Vargula luciferases
- Time-saving—assays using secreted luciferases require minimal sample handling
- Automation-friendly—amenable to high throughput screens
- Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
- Safe—allows one to perform non-radioactive assays
This Flash Assay Kit contains reagents for measuring the activity of Cypridina luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Cypridina Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Cypridina is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.
Includes:
Cell lysis buffer, reaction buffer and substrate
Requires:
Cypridina luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers.
Applications:
- Promoter studies for analyzing cis- regulatory elements and trans-acting factors
- Drug screening
- siRNA and miRNA screening
- Multiplexed assays to study off-target effects
- Secretory pathway/protein localization reporter assays
- Signal transduction pathway analysis
- RNA splicing studies
The Cypridina luciferase is a 61kDa protein from the marine ostracod, Cypridina noctiluca. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output captured using a luminometer can be correlated with the amount of Cypridina luciferase protein produced and used to determine the activity of the promoter driving Cypridina expression. The Cypridina gene in Thermo Scientific Cypridina Luc Vectors is modified for expression in mammalian cells.
The assay kit is optimized for use with our Cypridina Luc Vectors but is fully compatible with any luciferases that use Vargulin as a substrate. Because Cypridina uses a different substrate than Renilla, Gaussia, or firefly luciferases, co-transfection and assays with up to three reporters are possible. The signal produced by Cypridina shows strong flash kinetics and is considerably greater than flash signals from either firefly or Renilla luciferases assayed under similar conditions.
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Cypridina Luciferase |
| Quantity | 1000 Reaction Kit |
| Content And Storage | • Cypridina Flash Assay Buffer, 5 mL, store at 4°C • Vargulin (1X), 0.5 mL, store at -20°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature Upon receipt store kit at -20°C or store individual components as indicated above. |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer |
| Format | 96-well Plate, 384-well Plate |
| Label Type | Enzyme Labeled |
| Show More |
Frequently Asked Questions (FAQs)
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
Here are possible causes and solutions:
- Non-optimized lysis buffer used: Assay luciferase activity in the media to confirm good expression of luciferase; Use only the provided lysis buffer.
- Low luciferase expression: Lyse cells in smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale up volume of sample and reagent per well.
Here are possible causes and solutions:
- Insufficient luciferase accumulation in media: Incubate cells for a longer time.
- Low luciferase expression: Use less media per well during the experiment;.Use a different promoter or growth conditions to improve expression;.Increase the integration time on the instrument;.Scale-up the volume of sample and reagent per well.
- Treatment interfered with cellular secretory pathway: Transfect cells with a plasmid for constitutive expression of Luciferase (i.e., with pTK or pCMV promoter); Determine if luciferase actively expresses in media without treatment. Add treatment; Determine if there is a corresponding drop in luciferase activity from the constitutively expressed plasmid.
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
- No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
- Substrate auto-oxidized: Protect substrate from light and air; Maintain 100X Coelenterazine at -80 degrees C; maintain 100X Vargulin at -80 degrees C; Prepare new Coelenterazine Working Solution if used longer than 8 hours; Prepare new Vargulin Working Solution if used longer than 2 hours
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.