Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo Scientific™ iodoTMT Isobaric Label Reagents and Kits

Catalog No. p-4589202
Change view
Click to view available options
Quantity:
1 x 0.2 mg (per tag)
5 x 0.2 mg
5 x 0.2 mg (per tag)
Product Type:
Isobaric Label Reagent Set
Label Reagent
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PI90100 5 x 0.2 mg Label Reagent
PI90101 1 x 0.2 mg (per tag) Isobaric Label Reagent Set
PI90102 5 x 0.2 mg (per tag) Isobaric Label Reagent Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI90100 Supplier Thermo Scientific™ Supplier No. 90100
Only null left
Add to Cart
Add to Cart

Use Iodoacetyl Tandem Mass Tag (iodoTMT) Isobaric Label Reagents to label sulfhydryl-containing peptides for the multiplexed detection and quantitation of cysteine modification using tandem mass spectrometry analysis.

Thermo Scientific iodoTMT Label reagents enable multiplexed identification and quantitation of cysteine containing peptides in different samples using tandem mass spectrometry. IodoTMT reagents are sets of isobaric (mass and structure) isomers that are iodoacetyl-activated for covalent, irreversible labeling of sulfhydryl (–SH) groups. iodoTMTzero Label Reagent allows testing and optimization of sample preparation, labeling, fractionation, and MS fragmentation for peptide identification and reporter detection without using the more costly isotope-labeled compounds.

Iodoacetyl tandem mass tag reagents enable selective labeling of cysteine-containing peptides for multiplexed sample quantitation using LC-MS/MS analysis. Similar to iodoacetamide, iodoTMT reagents react specifically with reduced cysteines (Cys) in peptides and proteins and enable quantitation of the relative abundance of cysteine modifications, such as S-nitrosylation and the oxidation of disulfide bonds. IodoTMT reagents irreversibly label free sulfhydryl groups on cysteine residues, in contrast to previous reversible cysteine-reactive TMT (cysTMT) reagents.

To selectively analyze the cysteine-labeled peptides, the sample may be enriched using Immobilized Anti-TMT Antibody Resin (Cat. No. 90076) and TMT Elution Buffer (Cat. No. 90104). This approach of selective thiol-labeling and affinity enrichment is similar to isotope-coded affinity tags (ICAT method) but allows for affinity enrichment with a non-biological tag and higher multiplex quantitation.

Features of iodoTMT reagents
Specific—only reacts with sulfhydryl groups (reduced, unmodified cysteine residues)
Irreversible—labeled proteins and peptides are not susceptible to reducing agents
Flexible—options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
Versatile—can be used to study cysteine modifications (e.g., di-sulfide bridges, S-nitrosylation)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Label Reagent
Product Line iodoTMTzero
Content And Storage Store at –20°C.
Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 5 x 0.2 mg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.