Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Endotoxin-Free Tris Buffer, pH 7.0 (50 mM)

Catalog No. PIA43885 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA43885 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA43885 Supplier Thermo Scientific™ Supplier No. A43885
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pierce Endotoxin-Free Tris Buffer may be used to dilute samples for any of the endotoxin detection assays and for adjusting the pH of the LAL endotoxin test samples. For endotoxin testing, the samples and sample dilutions be between pH 6-8. Contains <0.005 EU/mL endotoxin.

Pierce Endotoxin-Free Tris Buffer can be used to dilute or adjust the pH of samples for endotoxin detection (LAL) assay. For endotoxin testing, the sample pH should be between pH 6-8. The buffer contains <0.005 EU/mL endotoxin.

TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1, 7732-18-5
Description Endotoxin-Free Tris Buffer
Concentration 50 mM
For Use With (Application) For adjusting the pH of the LAL endotoxin test samples
For Use With (Equipment) LAL Endotoxin Assay
pH 7
Physical Form Aqueous Solution
Quantity 5 x 10 mL
Solution Type Buffer
Packaging Vial(s)
Sterility Sterile
Endotoxin Concentration <0.005 EU/mL
Grade Biochemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.