Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluorescein-5-Maleimide

Catalog No. FER62245 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FER62245 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FER62245 Supplier Thermo Scientific™ Supplier No. 62245
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescein-5-Maleimide is a fluorescent reagent for labeling sulfhydryl-containing molecules.

Thermo Scientific™ FITC and Fluorescein Labeling Reagents and Kits are high-performance derivatives of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

  • Reacts with free sulfhydryl groups to form stable thioether bonds.
  • Requires application-specific optimization of the labeling protocol.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Maleimide
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C, protect from moisture
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes
Color Yellow to Orange
Product Line Pierce
Emission 518
Excitation 494
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.