Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Glutathione Superflow Agarose

Catalog No. PI25237 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI25237 50 mL
PI25236 10 mL
PI25238 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI25237 Supplier Thermo Scientific™ Supplier No. 25237
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.

Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.

Additional Information and Online Ordering

Pierce Glutathione Superflow Agarose is a highly crosslinked, durable resin that does not compress at flow rates required for medium- to large-scale FPLC purifications. The resin holds up well to a variety of chemical and pH values, and it is compatible with common clean-in-place procedures. The specific binding of glutathione to the GST-tag results in high purity and high yield in a single step purification process.

Highlights:

  • High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate

  • High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates

  • Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour

  • Compatible – maintains function after exposure to a wide variety of chemicals and pH values

  • Cost effective – resin is stable through multiple cycles of cleaning and reuse

Recommended for:

Medium- to large-scale FPLC purification of GST-tagged proteins

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at 4°C.
Product Line Pierce
Quantity 50 mL
Target GST-Tagged Protein
Type Agarose
Stationary Phase Glutathione
Format Bottle
Column Type Affinity, Agarose Resin
Binding Capacity 10 mg Pure GST Binding/mL Resin at 150 cm/hour Flow Rate, 20 mg/mL at 30 cm/hour Flow Rate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.