Learn More
Description
Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Additional Information and Online Ordering
Highlights:
High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate
High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour
Compatible – maintains function after exposure to a wide variety of chemicals and pH values
Cost effective – resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of GST-tagged proteins
Specifications
Specifications
| Capacity (Metric) | 10 mg Pure GST Binding per mL Resin at 150 cm/hour Flow Rate, 20 mg per mL at 30 cm/hour Flow Rate |
| Content And Storage | Upon receipt store at 4°C. |
| Product Line | Pierce |
| Quantity | 250 mL |
| Target | GST-Tagged Protein |
| Type | Agarose |
| Stationary Phase | Glutathione |
| Format | Bottle |
| Column Type | Affinity, Agarose Resin |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.