Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL

Catalog No. pi88274
Change view
Click to view available options
Quantity:
25 Columns
5 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88274 5 Columns
PI88275 25 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88274 Supplier Thermo Scientific™ Supplier No. 88274
Only null left
Add to Cart
Edge
Add to Cart

Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.

Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.

Highlights:

  • High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins

  • Durable re-use resin up to 10 times with no loss in performance

  • Selective recover at least 85% of your protein sample

  • High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL

  • Fast our new spin cup format enables endotoxin depletion within 1 hour

  • Cleansingle-use spin columns avoid cross contamination of samples

  • Optimized spin cups are optimized for different sample volumes

  • Economical large-volume discounts available

Requires:

Endotoxin-free water

Recommended for:

Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) Up to 2,000,000 EU/mL
Formulation 8mL-capacity centrifuge columns containing 0.5mL resin
Content And Storage Upon receipt store at 4°C.
Form Liquid
Product Line Pierce
Quantity 5 Columns
Target Endotoxin Removal
Type Spin Column
Stationary Phase Proprietary Ligand
Format Spin Column
Column Type Affinity, Cellulose Resin
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.