Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ High Capacity Protein A MagBeads, alkali stable

Catalog No. PIA53037 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
4 x 25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA53037 25 mL
PIA53035 1 mL
PIA53036 5 mL
PIA53038 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. PIA53037 Supplier Thermo Scientific™ Supplier No. A53037
Get 16% Off Protein Analysis Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Pierce High-Capacity Protein A MagBeads, Alkali Stable are a high-performance magnetic bead for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts.

Pierce High-Capacity Protein A MagBeads, Alkali Stable, are high-performance magnetic beads for purification of antibodies from cell culture supernatant, ascites, serum, and crude extracts.

Features of the High-Capacity Protein A MagBeads include:

  • High binding capacity—≥ 40 mg HIgG/mL settled beads
  • Alkaline stable—engineered to withstand standard CIP procedures with 0.1 M NaOH, enabling efficient cleaning of the beads for reuse
  • Animal component-free raw materials
  • Versatile—beads are compatible with manual and automated workflows (e.g., KingFisher instruments)
  • Stable—high-affinity Protein A is securely immobilized to magnetic agarose to help prevent leaching of ligand, target antibody, or base support

The protein A ligand has been genetically modified for stability in the presence of NaOH, so the beads can be regenerated at least 10 times without sacrificing binding capacity. The ligand is produced in yeast and is immobilized on magnetic agarose with an average particle size of ∼30 μm.

Pierce High-Capacity Protein A MagBeads, Alkali-Stable, are suitable for purifying mAbs and a subpopulation of scFv's and Fab fragments containing a VH3 domain. The beads are compatible with automation, allowing for higher throughput purifications and screening of purification conditions.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 6.25 mL
Binding Capacity ≥40 mg HIgG/mL settled beads
Capacity (Metric) 25 mL
Concentration Slurry: 25%
Description High Capacity Magnetic Agarose Beads
Content And Storage Provided in the form of a 25% slurry.

Upon receipt store at 2–8°C.

Particle Size 25 to 30 μm
Quantity 25 mL
For Use With (Application) Antibody Purification
Ligand Type Alkaline Stable Protein A
Format Bottle
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.