Learn More
Description
Thermo Scientific™ Pierce Protein A Plus UltraLink Resin is an extremely durable beaded polyacrylamide support to which IgG-binding Protein A has been immobilized at high density for use in IgG purification methods.
UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.
Highlights:
Immobilized Protein A - specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
High capacity - “Plus” variety has a dense load of immobilized Protein A, providing a binding capacity of at least 30mg human IgG per milliliter of resin
UltraLink Resin - acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Properties of Thermo Scientific UltraLink Resin:
- Support pH Stability: 1 to 13
- Average Particle Size: 50 to 80 microns
- Exclusion Limit: greater than 2,000,000 daltons
- Average Surface Area: greater than 250m2 per gram of beads
- Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)
- Pore Size: 500 to 1000 angstroms (50 to 100nm)
- Maximum Linear Velocity: 3000cm per hour
- Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
Specifications
Specifications
| Product Type | Resin |
| Product Line | Pierce, UltraLink |
| Column Type | UltraLink Affinity Resin |
| Format | Bottle |
| Stationary Phase | Protein A |
| Purification Target | IgG Antibodies |
| Content And Storage | Upon receipt store products at 4°C. Products are shipped at ambient temperature. |
| Quantity | 5 mL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.