Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip

Catalog No. PI15525 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15525 5 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15525 Supplier Thermo Scientific™ Supplier No. 15525

missing translation for 'validMainframeMsg'

Biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Thermo Scientific™ Pierce High Sensitivity Streptavidin Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Thermo Scientific Pierce Streptavidin Coated High Sensitivity Plates,Black, 8-Well Strip are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Features of Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip

• Sensitive—detect down to 5 ng/mL of biotinylated IgG
• Specific—low background with high signal-to-noise ratios
• Robust—broad dynamic range
• Ready to use—plates are supplied pre-blocked to save time

Thermo Scientific Pierce Coated Plates are coated using proprietary surface-activation technology to achieve maximum efficient coating of active streptavidin biotin-binding protein. The high sensitivity plates enable detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins.

Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low non-specific interactions.

Related Products
Pierce Streptavidin Coated High Sensitivity Plates, Clear, 8-Well Strip (Cat. No. 15520)

TRUSTED_SUSTAINABILITY

Specifications

Binding Property High Sensitivity, Affinity
Content And Storage Upon receipt store at 4°C. Product is shipped at ambient temperature.
Description Pierce Streptavidin Coated High Sensitivity Plates, Black, 96-Well Strip
Detection Method Fluorescence
Packaging 5 Plate
Product Line Pierce
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Streptavidin
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.