Learn More
Thermo Scientific™ MBP Column Preparation Buffer for Pierce™ IgM Purification Kit
Catalog No. PI21018 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
PI21018 50 mL
1 options
Catalog No. PI21018 Supplier Thermo Scientific™ Supplier No. 21018
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart
Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Thermo Scientific™ Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouse ascites fluid; the method uses immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5mL column of the MBP Agarose.
Highlights:
  • Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum
  • Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5mL ascites samples per column-use
  • Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies
  • Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity
Includes:
Kit contains a 5mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender
TRUSTED_SUSTAINABILITY
Specifications
Product Type IgM Purification Kit
Product Line Pierce
Reagent Type Conditioning Reagent
Description Pierce MBP Column Preparation Buffer
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C.
Quantity 50 mL
Form Liquid
For Research Use Only. Not for use in diagnostic procedures.