Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immobilized Pepsin (Agarose Resin)

Catalog No. PI20343 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20343 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20343 Supplier Thermo Scientific™ Supplier No. 20343
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Generate and purify Fab and F(ab')2 fragments from antibodies with acidic endopeptidase (pepsin) immobilized onto beaded agarose resin.

Thermo Scientific™ Pierce Immobilized Pepsin consists of an acidic endopeptidase (pepsin) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab and F(ab')2 fragments from antibodies.

Immobilized pepsin can be substituted for free pepsin in any application, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the pepsin. Immobilized pepsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity. The F(ab')2 Preparation Kit contains immobilized pepsin, other necessary reagents and an optimized protocol for human, rabbit and mouse antibody digestion.

Highlights:

  • Pepsin can cleave Fc fragments from IgG, resulting in F(ab')2 fragments
  • Broad specificity, including esterase activity
  • Easy to use - simply remove the agarose beads to stop the enzymatic digestion

Includes:

Kit contains immobilized pepsin, other necessary reagents and an optimized protocol for human, rabbit and mouse antibody digestion.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store product at 4-8°C. Product is shipped at ambient temperature.
Form Liquid Suspension
Quantity 5 mL
Product Type Pepsin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.