Learn More
Description
Thermo Scientific™ Pierce Immobilized Reductant Column contains beaded resin to which an thiol-based reducing agent has been immobilized to enable solid-phase reduction of peptide and protein disulfide bonds.
Highlights:
- Eliminates soluble components which interfere with the assay of free thiol groups
- No need to remove reducing agent after the reaction
- Reduced protein, free of contaminating thiols, can be easily assayed using the BCA method
- Solid phase; protocols for both peptides and proteins
- Excellent protein recovery
- Regenerable matrix usable up to 4 times
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. |
| Product Line | Pierce |
| Form | Liquid |
| Quantity | 1 column |
| Product Type | Reductant Column |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.