Learn More
Thermo Scientific™ Pierce™ ImmunoPure™ Polyclonal Goat Anti-Hamster IgG (H+L) Purified Antibodies
Shop All Fisherbrand ProductsDescription
- F(ab)2 fragments of antibodies are especially useful in assays in which binding between the Fc portions of antibodies and Fc receptor bearing cells must be eliminated
- Purified by immunoaffinity chromatography using antigens coupled to agarose gels, resulting in an increase in sensitivity and specificity, with a decrease in background and yielding antibodies with high avidity
- Exhibit both maximal binding to antigens and minimal cross-reactivity to other molecules
- Conjugated antibodies are affinity-purified prior to the conjugation process
- Selected ImmunoPure™ antibodies have been further purified by passing them through immobilized serum proteins from other species
- Additional processing minimizes cross-reactivities to other species' serum proteins
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Host Species | Goat |
| Quantity | 1.5 mg |
| Primary or Secondary | Secondary |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.