Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Inclusion Body Solubilization Reagent

Catalog No. PI78115 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78115 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI78115 Supplier Thermo Scientific™ Supplier No. 78115
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Retrieve recombinant proteins that form insoluble aggregates during bacterial overexpression with this reagent optimized for inclusion body solubilization.

Effectively retrieve recombinant proteins that form insoluble aggregates during bacterial overexpression with Thermo Scientific™ Inclusion Body Solubilization Reagent.

The Inclusion Body Solubilization Reagent is a ready-to-use aqueous solution of denaturant that helps to dissolve aggregated proteins (inclusion bodies) that have been separated from the soluble fraction of bacterial cell lysates. The reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents such as Thermo Scientific B-PER Bacterial Protein Extraction Reagent. Bacteria are widely used for recombinant protein expression. However, 70%-80% of proteins produced by recombinant techniques in E. coli form inclusion bodies (i.e., protein aggregates). Once these aggregates are formed, it is very difficult to solubilize them. A proprietary denaturant contained in this reagent provides the most effective means for solubilizing aggregated proteins. Additional components, such as a reducing agent and a chelating agent, may be added to the reagent, depending on the particular application.

Highlights:

  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy- and ready-to-use solution; no preparation needed
  • Compatible with Thermo Scientific Coomassie Plus Protein Assay Kit (Part No. 23236) to determine degree of expression
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE can be performed
  • Can accommodate additional components such as reducing agents or chelating agents

Compatible with:

Thermo Scientific Pierce Protein Refolding Kit (Part No. 89867)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Solubilization Reagent
Content And Storage Upon receipt store at room temperature.
Form Liquid
Reagent Type Chaotrope, Denaturant
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.