Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)

Catalog No. PIA40010 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
0.5 pmol/μL
5 pmol/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
PIA40010 0.5 pmol/μL
FERA51747 5 pmol/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA40010 Supplier Thermo Scientific™ Supplier No. A40010

missing translation for 'validMainframeMsg'

Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven HeavyPeptide AQUA Ultimate peptides provided at five dilutions distinguished by differential isotopic labeling to assess sensitivity and dynamic range of LC-MS/MS systems.

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mix) contains 7 of 15 peptides from the Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88320). Each of the seven peptides has five versions (amino acids labeled with 0, 1, 2, 3, or 4 heavy isotopes) at five distinct concentrations. The 7 x 5 mix can be used with Skyline software to assess dynamic range, linearity, and lower limit of quantitation (LLOQ).

The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
• High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
• Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
• Quantitative—assesses dynamic range of instrument and instrument sensitivity
• Predictive—enables assessment of chromatography and MS instrument performance

Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) (0.5 pmol/μL)
Product Line Pierce
Shipping Condition Dry Ice
Concentration 0.5 pmol/μL
For Use With (Equipment) Mass Spectrometer
Type LC-MS/MS System Standard
Quantity 25 μL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.