Learn More
Description
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
Highlights:
- Highly purified, recrystallized reagent
- Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
- Provides accurate quantity of matrix every time
Specifications
Spécifications
| Description | Pierce SA MALDI Matrices |
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | MALDI Matrix |
| Content And Storage | Upon receipt store product at 4°C. |
| Starting Material | Small Molecules |
| Final Product Type | Small Molecules |
| Detection Method | Mass Spectrometry |
| Workflow Step | Calibration |
| Quantity | 24 x 1 mg |
| Form | Solid |
Sécurité et manipulation
For Research Use Only. Not for use in diagnostic procedures.
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.