Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Maleimide Activated Horseradish Peroxidase

Catalog No. PI31485 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31485 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31485 Supplier Thermo Scientific™ Supplier No. 31485
  • PRODUCT PROMOTION
    $487.00
    $313.14 / Pack of 1

    Save $173.86

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activated horseradish peroxidase (HRP) enzyme and kit to label proteins, peptides or other ligands via sulfhydryl groups (reduced cysteines) to make HRP conjugates.

Thermo Scientific™ Pierce Maleimide-Activated HRP Conjugation Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.

The Thermo Scientific Maleimide Activated Horseradish Peroxidase (HRP) Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain a free sulfhydryl group. This kit eliminates the first step of the two-step maleimide method, making conjugate preparation much simpler. The kit is supplied with a stable, Maleimide-activated horseradish peroxidase. HRP conjugates can be purified by gel filtration chromatography, ultrafiltration or dialysis, depending upon the size of the conjugated protein.

Highlights:

  • Prepare enzyme conjugates from proteins that contain a free sulfhydryl
  • Includes 2-MEA to generate free sulfhydryls from disulfide bonds
  • Includes SATA to add free sulfhydryls to lysine residues
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Thiol
Content And Storage Note: Total weight is ∼25mg as a result of buffer components and salts lyophilized along with the activated protein

Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Formulation Activated horseradish peroxidase lyophilized from PBS
Form Solid
Product Line EZ-Link
Label Type Enzyme Labeled
Label or Dye HRP (Horseradish Peroxidase)
Reactive Moiety Maleimide
Solubility Water
Quantity 5 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.