Learn More
Description
Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.
Highlights:
- Pure compounds of specific size ensure complete control of PEGylation
- Straight-chain compounds with simple terminal primary amine
- Adaptable to a variety of modification procedures
Recommended for:
- PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
Specifications
Specifications
| Chemical Reactivity | Ketone |
| Description | Pierce MA(PEG)24 |
| Content And Storage | Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature. |
| Label Type | Polyethylene Glycol (PEG), Blocking Groups |
| Reactive Moiety | Amine, Primary Amine |
| Spacer | 24 ethylene glycol units |
| Form | Solid |
| Quantity | 1 g |
| Solubility | Water |
| Product Type | Amine Compound |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.