Learn More
Description
Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.
Highlights:
- NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Easy-to-follow instructions increase the likelihood of a successful outcome
Specifications
Specifications
| Chemical Reactivity | Amine |
| Content And Storage | Store desiccated at -20°C. |
| Label Type | Polyethylene Glycol (PEG), Blocking Groups |
| Product Line | Pierce |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Spacer | 4 ethylene glycol units |
| Form | Solid |
| Quantity | 1 g |
| Solubility | Water |
| Product Type | NHS-Ester Reagent |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.