Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Micrococcal Nuclease Solution (≥100 U/μL)
GSA_VA

Catalog No. PI88216 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88216 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88216 Supplier Thermo Scientific™ Supplier No. 88216
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Remove nucleic acids from cell lysates, release chromatin-bound proteins and shear chromatin for chromatin immunoprecipitation (ChIP) using micrococcal nuclease.

Thermo Scientific™ Micrococcal Nuclease (MNase) is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.

This micrococcal nuclease is a stable liquid form of the enzyme derived from Staphylococcus aureus. Micrococcal nuclease (MNase) exhibits exo- and endo-5'-phosphodiesterase activities against DNA and RNA. This enzyme digests double-stranded, single-stranded, circular and linear nucleic acids. The highest activity is toward single-stranded nucleic acid substrates with preference for AT- or AU-rich regions. Enzymatic activity occurs at pH 7 to 10 and is strictly dependent on calcium for the digestion of RNA and DNA substrates. Micrococcal nuclease is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.

Highlights:

  • Digests nucleic acids (DNA and RNA)

  • Effective for double-stranded, single-stranded, circular and linear nucleic acids
  • Active in neutral to alkaline buffer conditions containing divalent calcium ions
  • Supplied at ≥100 units/μL in 10mM Tris-HCl, pH 7.5, 50mM NaCl, 1mM EDTA, 50% glycerol

Compatible with:

Thermo Scientific Subcellular Protein Fractionation Kit (Part No. 78840); Thermo Scientific Pierce ChIP Kit (Part No. 26156)

Recommended for:

Enzymatic DNA shearing for chromatin immunoprecipitation assays (ChIP); Removal of unwanted genomic or organellar nucleic acids from cell lysates; Degradation of nucleic acids in subcellular fractionation procedures

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Upon receipt store at -20°C. Product is shipped with dry ice.
Form Liquid
Reagent Type Enzyme for Cell Lysis
Quantity 150 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.