Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Mini Gel Power Staining Kit

Catalog No. pi22840
Change view
Click to view available options
Quantity:
30 gels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22840 30 gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22840 Supplier Thermo Scientific™ Supplier No. 22840
Only null left
Add to Cart
Add to Cart

The Thermo Scientific Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Mini Gel Power Staining Kit enables rapid Coomassie dye staining of proteins mini-size polyacrylamide gels and the removal of unbound stain from the gel matrix.

The Thermo Scientific Pierce Power Staining kits are designed for fast Coomassie dye staining of protein gels. The Pierce Mini Gel Power Staining Kit enables rapid Coomassie dye staining of proteins mini-size polyacrylamide gels and the removal of unbound stain from the gel matrix. Two mini gels having the same formulation can be accommodated and stained simultaneous in the same power stain cassette.

  • High performance—performance is equivalent to, or better than, traditional Coomassie dye staining techniques
  • Fast—optimized ionic solutions enable rapid Coomassie dye staining of protein and gel destaining.
  • Compatible—verified to work with commonly used precast and homemade SDS-PAGE gels

Traditional Coomassie dye staining techniques require staining for one hour to overnight and prolonged destaining for desired results. When used together, the Pierce Power stain solution, destain solution, and gel pads enable efficient staining and destaining in minutes with results equivalent to, or better than, traditional Coomassie dye staining techniques.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Note: The kit contains sufficient reagents to stain 30 mini-sized gels at 15 mL each.

Contents:
Mini Staining Gel Pads, 2 boxes of 30 each (8 sheets/pad)
Pierce Power Stain Solution, 480 mL
Destain Solution, 480 mL
Description Mini Gel Power Staining Kit, 30 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 30 Mini-Sized Gels at 15mL Each
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye Coomassie
Formulation Proprietary gel stain and destain solutions
Quantity 30 gels
Sufficient For 30 mini-sized gels at 15mL each
Target Molecule Protein
Product Line Pierce
Product Type Fast Protein Gel Stain
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.