Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ MMTS (methyl methanethiosulfonate)

Catalog No. PI23011 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23011 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23011 Supplier Thermo Scientific™ Supplier No. 23011
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Methyl methanethiosulfonate (CAS 2949-92-0); reversibly blocks cysteines (sulfhydryl groups) to facilitate study of enzyme activation and protein functions.

Thermo Scientific™ Pierce MMTS is methyl methanethiosulfonate, a small compound that enables cysteines and other sulfhydryl groups to be reversibly blocked to facilitate study of enzyme activation and other protein functions.

Highlights:

  • Converts -SH groups to -S-S-CH3
  • Liquid with density of 1.337 (10 M, ∽150 μL)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Thiol
Content And Storage Upon receipt store at 4°C.
Label or Dye Unreactive Group
Label Type Blocking Groups
Product Line Pierce
Reactive Moiety Thiol
Form Liquid
Quantity 200 mg
Product Type Methyl Methanethiosulfonate
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.