Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Monomeric Avidin UltraLink™ Resin

Catalog No. PI53146 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI53146 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI53146 Supplier Thermo Scientific™ Supplier No. 53146
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reversibly bind biotinylated proteins and other ligands with monomers of avidin covalently attached as individual subunits onto beaded resins.

Thermo Scientific™ Pierce Monomeric Avidin Resins and Kit enable gentle, reversible-binding and purification of biotinylated proteins.

Pierce Monomeric Avidin Kit and Resins are unique among biotin-binding supports in allowing non-denaturing affinity purification of biotinylated molecules. The immobilized monomeric avidin protein binds biotin with high specificity and moderate affinity, allowing non-biotin molecules to be washed away and then the bound biotin-labeled molecules to be competitively eluted using 2mM biotin in phosphate buffered saline (PBS). This technique provides the gentlest elution conditions for biotinylated protein purification and avoids the contamination and other problems associated with traditional avidin and streptavidin methods. The monomeric avidin column can then be regenerated by using 0.1M glycine to strip the column of residual bound biotin without losing the ability to bind another biotinylated sample.

With typical avidin or streptavidin, the biotin-binding affinity (Kd = 10-15M) is so great that purification with these traditional media require denaturing conditions for elution, such as 8M Guanidine·HCl at pH 1.5 or boiling in reducing SDS-PAGE sample loading buffer. In addition to their adverse effects on the biotinylated protein of interest, such harsh elution conditions typically cause extraneous proteins (i.e., the avidin or streptavidin subunits) to strip from the resin and co-elute with the desired purification product. By contrast, when avidin is coupled to a resin (e.g., crosslinked beaded agarose or UltraLink Resin) as the subunit monomer, its specificity for biotin is retained, but its biotin-binding affinity decreases to a level (Kd = 10-8M ) that is conducive to subsequent elution.

Highlights:

  • Non-denaturing – purifies biotinylated products using mild elution conditions (2mM free biotin)

  • Reusable – Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)

  • Specific – retains biotin-binding specificity and low nonspecific binding of native avidin

  • Flexible – bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2mM biotin

  • Good binding capacity – greater than 1.2mg biotinylated BSA/mL of resin

  • Convenient – purchase standalone agarose or UltraLink™ Resins or a complete purification kit with prepacked column and buffers

Item-Specific Features:

  • Agarose Resins and Kit are based on crosslinked 4% beaded agarose
  • UltraLink Resin is an acrylamide support (UltraLink Biosupport)

Includes:

Kit contains a pre-packed column of Monomeric Avidin Agarose and binding and elution buffers.

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Monomeric Avidin UltraLink Resin
Content And Storage Upon receipt store at 4°C.
Product Line Pierce, UltraLink
Quantity 5 mL
Target Biotin-Tagged Molecule
Type Resin
Stationary Phase Avidin
Format Bottle
Column Type Affinity, UltraLink™ Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.