Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mouse Embryonic Fibroblast Isolation Enzyme 1 (with papain) for Pierce™ Primary Cell Isolation Kits

Catalog No. PI88290 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88290 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88290 Supplier Thermo Scientific™ Supplier No. 88290
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ensure optimal yield and viability of fibroblasts from mouse embryos with the isolation and culturing reagents in this kit.

Ensure optimal yield and viability of fibroblasts from mouse embryos with the isolation and culturing reagents in the Thermo Scientific™ Pierce™ Mouse Embryonic Fibroblast Isolation Kit.

The Thermo Scientific Pierce Mouse Embryonic Fibroblast Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of fibroblasts from mouse embryos.

The Mouse Embryonic Fibroblast Isolation Kit has been designed to obtain high viability and purity of embryonic fibroblasts using validated reagents and an optimized procedure. The kit contains a tissue-specific dissociation enzyme and media formulated for primary cell culture that promotes the isolation growth of functional fibroblasts. In addition, MEF isolated and cultured with Pierce MEF Isolation Kit can be cryopreserved for future use with very minimal loss of viability.

Highlights:
  • Optimized – procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Yield – provides a 2-fold increase in yield compared to do-it-yourself methods and other commercial kits
  • Functional – cultured fibroblasts express the appropriate biochemical markers
Includes:
  • Complete kit includes Fibroblast Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), and DMEM for Primary Cell Isolation
Applications:
  • Generation of immortalized cell lines from transgenic mice
  • Feeder layer for stem cells
  • IHC and IF studies
Product Details:

Because it uses an optimized tissue-specific dissociation enzyme, this kit isolates fibroblasts with greater yield and cell viability than cells isolated by existing methods. Isolated MEF express fibroblast-specific protein markers, confirming their viability and unperturbed metabolic function. After isolation, the mouse fibroblasts may be immortalized by continuous passage for the establishment of unique cell lines. They may also be used as a cell culture model for a diverse range of studies such as gene regulation and stem cell research [1,2,3,4].

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Isolation Kit
Content And Storage Upon receipt store product at 2-80°C. Product is shipped on ice packs.
Cell Type Fibroblasts
Target Species Mouse
Form Solid
Product Line Pierce
Quantity 1 Vial
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.