Learn More
Thermo Scientific™ Pierce™ NHS-Activated Agarose Spin Columns, 0.2 mL
Shop All Thermo Scientific ProductsDescription
Perform rapid and stable immobilization of proteins, peptides and other ligands with Thermo Scientific™ Pierce NHS-Activated Agarose, a high-quality beaded-agarose support.
The Pierce NHS-Activated Agarose resin is available in an anhydrous acetone slurry and as dry powder. The unique dry form does not require storage in or removal and disposal of the acetone solvent. In addition, the dry resin is ideal for coupling reactions with dilute samples because it concentrates the sample as the resin swells, reducing the volume of the starting material and resulting in highly efficient ligand immobilization.
Highlights:
Easy to use – immobilize in a simple one-step reaction with minimal hands-on time
Rapid and efficient – greater than 85% coupling for most proteins within 30 minutes
Innovative – the dry agarose concentrates the sample, making it ideal for immobilizing dilute proteins
Safe – no hazardous chemicals needed (e.g., sodium cyanoborohydride, cyanogen bromide)
Versatile – affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications
Compatible – use with any primary amine-containing compound
Reusable – the leak-resistant chemistry means you can reuse the affinity resin
High binding capacity – binding capacity of >30mg/mL (slurry) or >25mg/mL (dry resin)
Specifications
Specifications
| Content And Storage | Upon receipt store dry resin at -20°C. Product is shipped in an ice pack. |
| Form | Liquid |
| Product Line | Pierce |
| Quantity | 25 Columns |
| For Use With (Application) | Affinity Chromatography |
| Protein Form | Recombinant |
| Target | Antibody, Protein, Peptide, Ligand |
| Type | Spin Column |
| Stationary Phase | NHS Ester |
| Sufficient For | ≥25 mg per mL Dry Resin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.