Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ NHS-Activated Agarose Slurry

Catalog No. PI26200 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26200 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26200 Supplier Thermo Scientific™ Supplier No. 26200
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

N-Hydroxysuccinimide ester resin and columns to covalently immobilize proteins, peptides and other amine-containing ligands for affinity purification.

Perform rapid and stable immobilization of proteins, peptides and other ligands with Thermo Scientific™ Pierce NHS-Activated Agarose, a high-quality beaded-agarose support.

The activated affinity resin is available as a slurry or dry powder, allowing simple and efficient immobilization of proteins to a beaded-agarose support. The beaded agarose support contains N-hydroxysuccinimide (NHS) functional groups that react with primary amines to form stable amide linkages. The coupling reaction is performed in an amine-free buffer at pH 7 to 9. Protein coupling efficiency is typically greater than 80%, regardless of the molecular weight or pI the ligand. Coupling capacity exceeds 25mg of protein per milliliter of resin. Once a ligand is immobilized, the prepared resin can be used for multiple affinity purification procedures. The crosslinked beaded agarose has fast linear flow potential, making it useful for gravity-flow and low- to medium-pressure applications.

The Pierce NHS-Activated Agarose resin is available in an anhydrous acetone slurry and as dry powder. The unique dry form does not require storage in or removal and disposal of the acetone solvent. In addition, the dry resin is ideal for coupling reactions with dilute samples because it concentrates the sample as the resin swells, reducing the volume of the starting material and resulting in highly efficient ligand immobilization.

Highlights:

  • Easy to use – immobilize in a simple one-step reaction with minimal hands-on time

  • Rapid and efficient – greater than 85% coupling for most proteins within 30 minutes

  • Innovative – the dry agarose concentrates the sample, making it ideal for immobilizing dilute proteins

  • Safe – no hazardous chemicals needed (e.g., sodium cyanoborohydride, cyanogen bromide)

  • Versatile – affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications

  • Compatible – use with any primary amine-containing compound

  • Reusable – the leak-resistant chemistry means you can reuse the affinity resin

  • High binding capacity – binding capacity of >30mg/mL (slurry) or >25mg/mL (dry resin)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store the agarose slurry at 4°C. Product is shipped at ambient temperature.
Form Liquid
Product Line Pierce
Quantity 25 mL
Protein Form Recombinant
Target Antibody, Protein, Peptide, Ligand
Type Agarose
Stationary Phase NHS Ester
Format Bottle
Column Type Affinity, Agarose Resin
Binding Capacity ≥30 mg/mL Coupling
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.