Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lysosome Enrichment Kit for Tissues and Cultured Cells

Catalog No. PI89839 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
230 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89839 230 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89839 Supplier Thermo Scientific™ Supplier No. 89839
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Enrich for lysosomes, peroxisomes and nuclei from cells and tissue with these organelle-specific subcellular fractionation kits.

Perform enrichment of intact organelles including lysosomes, peroxisomes and nuclei from cells and tissue with Thermo Scientific™ Organelle Enrichment Kits.

These three organelle-specific kits provide the media and reagents necessary to accomplish subcellular fractionation using high-speed density gradient centrifugation. Subcellular fractionation simplifies otherwise complex protein mixtures that result from whole cell lysis, thereby facilitating proteomic analysis of specific organelle targets and pathways. Each kit uses density gradient centrifugation to separate organelles from contaminating cellular structures. The isolated organelles may be used for a number of downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localization studies.

Highlights:

  • Efficient and easy to use – kit reagents and gradient centrifugation separate organelles from contaminating structures

  • Compatible – prepare samples for downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localization studies

  • Complete – kits contain sufficient material for 25 applications

Includes:

Organelle-specific Reagents A and B, Optiprep™ Cell Separation Media, and phosphate-buffered saline

Requires:

Ultracentrifuge, refrigerated microcentrifuge, vortex mixer, Dounce tissue grinder, sonicator

TRUSTED_SUSTAINABILITY

Specifications

Product Type Lysosome Enrichment Kit
Content And Storage Upon receipt store at 4°C. Product shipped at ambient temperature
Form Liquid
Reagent Type Organelle Isolation, Subcellular Fractionation
Sufficient For 25 Samples of 50 to 200 mg Cells or Tissues Each
Quantity 230 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.