Learn More
Description
Thermo Scientific™ Pierce™ Peptide Clean-Up Plate products are sorbent-based 96-well plates designed for parallel peptide desalting, clean-up, concentration, and fractionation prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows. These plates help remove salts, detergents, buffers, denaturants, chaotropes, and other chemistry-compatible contaminants that can interfere with downstream analysis. The plate portfolio includes multiple peptide clean-up chemistries, including octadecyl silica (C18), strong anion exchange (SAX), mixed-mode cation exchange (MCX), styrene divinylbenzene reversed-phase sulfonate (SDB-RPS), and styrene divinylbenzene crosslinked (SDB-XC), for high-throughput desalting, clean-up, concentration, and fractionation workflows.
Thermo Scientific™ Pierce™ Peptide Clean-Up Plate products use reversed-phase, ion-exchange, or mixed-mode retention to support a range of peptide sample-preparation strategies. The 96-well plate format can be used in centrifugation-based or automation-compatible workflows where appropriate for the selected chemistry and protocol.
Features of Thermo Scientific™ Pierce™ Peptide Clean-Up Plate, 96 well
- High-throughput - process up to 96 peptide samples in a plate-based format
- Flexible - choose from multiple chemistries to support plate-based peptide clean-up, desalting, concentration, and fractionation workflows
- Efficient - help remove LC-MS-incompatible contaminants and prepare peptide samples in MS-compatible solutions for downstream analysis
- Automation-compatible - support automated sample-preparation workflows where appropriate for the selected chemistry and protocol
Applications
- High-throughput peptide desalting prior to liquid chromatography-mass spectrometry (LC-MS) analysis
- Plate-based clean-up following enzymatic protein digestion workflows
- Removal of chemistry-compatible salts, buffers, detergents, denaturants, chaotropes, lipids, and other liquid chromatography-mass spectrometry (LC-MS) interferences
- Plate-based peptide concentration and mass spectrometry sample preparation
- Peptide fractionation to reduce sample complexity and support deeper proteome analysis
Specifications
Specifications
| Product Type | Spin Plate |
| Product Line | Pierce |
| Starting Material | Peptides, Protease-digested Protein |
| Detection Method | Mass Spectrometry |
| Format | Plate |
| Quantity | 1 x 96-well Plate |
| Form | Solid |
| Sorbent | SDB-XC |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.