Learn More
Description
Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips are sorbent-based devices designed for rapid, reproducible peptide desalting, clean-up, and concentration prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows. These spin tips enable efficient peptide binding and recovery while removing salts, detergents, buffers, denaturants, chaotropes, and other chemistry-compatible contaminants that can interfere with downstream analysis. Available chemistries include octadecyl silica (C18), strong anion exchange (SAX), strong cation exchange (SCX), mixed-mode cation exchange (MCX), styrene divinylbenzene reversed-phase sulfonate (SDB-RPS), and styrene divinylbenzene crosslinked (SDB-XC), enabling selection based on the sample matrix, contaminant profile, and desired clean-up or fractionation strategy.
When used with Pierce™ Spin Tip adaptors (Catalog No. A40011776) and low protein binding microcentrifuge tubes (Catalog No. 90410), Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips support a simple centrifugation-based workflow with standard laboratory equipment. The spin tip format provides a straightforward setup for peptide desalting, clean-up, concentration, or chemistry-compatible fractionation before downstream MS analysis.
Features of Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips
- Efficient - help remove interfering contaminants to improve spectral quality and prepare peptide samples in mass spectrometry (MS)-compatible solutions
- Flexible - choose from multiple chemistries to support peptide desalting, clean-up, concentration, and fractionation strategies
- Convenient - process samples by standard centrifugation without specialized equipment using Pierce Spin Tip adaptors and low protein binding microcentrifuge tubes
Applications
- Peptide desalting prior to liquid chromatography-mass spectrometry (LC-MS) analysis
- Clean-up following enzymatic protein digestion workflows
- Removal of chemistry-compatible salts, buffers, detergents, denaturants, chaotropes, lipids, and other liquid chromatography-mass spectrometry (LC-MS) interferences
- Peptide concentration and mass spectrometry sample preparation
- Peptide fractionation to reduce sample complexity and support deeper proteome analysis
Specifications
Specifications
| Product Type | Spin Tip |
| Product Line | Pierce |
| Starting Material | Peptides, Protease-digested Protein |
| Detection Method | Mass Spectrometry |
| Format | Tip |
| Workflow Step | Peptide Clean-up |
| Quantity | 96 Tips |
| Form | Solid |
| Sorbent | SDB-RPS |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.