Learn More
Description
Thermo Scientific™ Pierce Peroxidase Conjugate Stabilizer is an antifreeze solution for storing aliquots and stocks of antibodies and other HRP-conjugates so they can be pipetted for use without thawing.
Highlights:
- Provides a buffered antifreeze environment for enzyme conjugated antibodies and proteins Maintains a liquid, low viscosity solution at -20°C
- No aliquoting of conjugate is necessary and sampling is convenient
- Contains preservatives to prevent anti-microbial growth during long-term storage
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. |
| Form | Liquid |
| Product Line | Pierce |
| Reagent Type | Stabilizing Reagent |
| Validated Application | ELISA, Immunohistochemistry, Western Blot |
| Quantity | 25 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.