Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Phosphoprotein Enrichment Kit
GSA_VA

Catalog No. pi90003
Change view
Click to view available options
Quantity:
10 Column Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI90003 10 Column Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI90003 Supplier Thermo Scientific™ Supplier No. 90003
Only null left
Add to Cart
Add to Cart

Purify (enrich) phosphorylated proteins from mammalian cell and tissue lysates using phospho-specific IMAC affinity resin cartidges or kit.

Perform efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues with Thermo Scientific™ Pierce Phosphoprotein Enrichment Kit.

The kit is an effective system for isolating the phosphoprotein fraction of mammalian cell and tissue lysates. The included lysis-binding buffer and proprietary affinity resin produce superior yields with negligible non-specific binding. The kit uses a convenient spin column format, and the resulting phosphorylated protein sample is compatible with downstream applications, including Western blotting, mass spectrometry (MS), 2D-PAGE, and protein arrays. The affinity resin is also available as pre-packed chromatography cartridges for use with automated liquid chromatography (LC) systems.

Highlights:

  • Specific – low nonspecific protein contamination from complex biological samples, such as cell culture lysate and mouse tissue extract

  • Fast – easy spin format enables enrichment of phosphorylated proteins in less than 2 hours

  • Superior yield – achieves higher yields than other commercially available phosphoprotein isolation kits

  • Convenient format – complete kit offering pre-dispensed spin columns, buffers, reagents and ultrafiltration columns for enrichment

  • Compatible – downstream applications include mass spectrometry, Western blotting and 2D-PAGE

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at 4°C.
Product Line Pierce
Quantity 10 Column Kit
Target Phosphoprotein
Type Phosphoprotein Enrichment Kit
Stationary Phase Proprietary Ligand
Sufficient For 10 Reactions
Format Spin Column
Column Type Affinity, Agarose Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.